missing translation for 'onlineSavingsMsg'
Learn More

3-morpholinobenzaldehyde, 97%, Thermo Scientific™

Catalog No. CC33804CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC33804CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC33804CB Supplier Thermo Scientific Chemicals Supplier No. CC33804CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446866-87-1
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
InChI Key LQORKFSMUOSSQM-UHFFFAOYSA-N
Synonym 3-morpholinobenzaldehyde, 3-morpholin-4-yl benzaldehyde, 4-3-formylphenyl morpholine, 3-4-morpholinyl benzaldehyde, 3-morpholin-4-yl-benzaldehyde, pubchem13334, 3-morpholin-4-yl benzaldehyd, benzaldehyde,3-4-morpholinyl
PubChem CID 7164584
IUPAC Name 3-morpholin-4-ylbenzaldehyde
SMILES C1COCCN1C2=CC=CC(=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.23
Percent Purity 97%
Chemical Name or Material 3-Morpholinobenzaldehyde