accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 4437-51-8

IUPAC Name: hexane-3,4-dione
Synonyms: 3,4-hexanedione
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: KVFQMAZOBTXCAZ-UHFFFAOYSA-N
SMILES: CCC(=O)C(=O)CC
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)