missing translation for 'onlineSavingsMsg'
Learn More

3,4-Hexanedione 96,0+%, TCI America™

Numéro de catalogue. H05735ML
Modifier l'affichage
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H05735ML 5 mL
H057325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H05735ML Fournisseur TCI America Code fournisseur H05735ML

Identifiants chimiques

CAS 4437-51-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00010237
InChI Key KVFQMAZOBTXCAZ-UHFFFAOYSA-N
Synonym 3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
PubChem CID 62539
IUPAC Name Hexane-3,4-dione
SMILES CCC(=O)C(=O)CC

Spécifications

Color Jaune
Boiling Point 130°C
Quantity 5 mL
UN Number 1224
Formula Weight 114.14
Percent Purity ≥96.0% (GC)
Physical Form Liquide
Chemical Name or Material 3,4-Hexanedione
TSCA Yes
Recommended Storage Congélateur