accessibility menu, dialog, popup

UserName

CAS RN 442633-00-3

CH3HNNHOF

CAS RN 442633-00-3

IUPAC Name: 2-fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]benzamide
Synonyms: 2-fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]benzamide
Molecular Weight (g/mol): 296.35
Molecular Formula: C18H17FN2O
InChi Key: UXRKUKRXVWJFER-UHFFFAOYSA-N
SMILES: CC1=C(CCNC(=O)C2=CC=CC=C2F)C2=CC=CC=C2N1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results