missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Arp2/3 Complex Inhibitor I, CK-666

Catalog No. 18251525MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18251525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18251525MG Supplier MilliporeSigma™ Supplier No. 18251525MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Arp2/3 Complex Inhibitor I, CK-666, also referenced under CAS 442633-00-3, controls the biological activity of Arp2/3. This small molecule/inhibitor is primarily used for Cell Structure applications.

Packaged Under Inert Gas

Specifications

Color Off-white
Description 95% by HPLC
Product Type Inhibitor
CAS 442633-00-3
Molecular Formula C18H17FN2O
Formula Weight 296.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg