accessibility menu, dialog, popup

UserName

CAS RN 4407-36-7

IUPAC Name: (2E)-3-phenylprop-2-en-1-ol
Synonyms: cinnamyl alcohol
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: OOCCDEMITAIZTP-QPJJXVBHSA-N
SMILES: OC\C=C\C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results