missing translation for 'onlineSavingsMsg'
Learn More

(E)-Cinnamyl Alcohol 97.0+%, TCI America™

Catalog No. C0362500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0362500G 500 g
C036225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0362500G Supplier TCI America Supplier No. C0362500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4407-36-7
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002921
InChI Key OOCCDEMITAIZTP-QPJJXVBHSA-N
Synonym (E)-3-Phenyl-2-propen-1-ol
PubChem CID 5315892
ChEBI CHEBI:33227
IUPAC Name (E)-3-phenylprop-2-en-1-ol
SMILES C1=CC=C(C=C1)C=CCO

Specifications

Color White-Yellow
Boiling Point 250°C
Quantity 500 g
Formula Weight 134.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (E)-Cinnamyl Alcohol
EINECSNumber (3)-1039
RTECSNumber GE2200000
TSCA Yes