accessibility menu, dialog, popup

UserName

OHONNHClOHSOHOO(Z)

CAS RN 4386-25-8

IUPAC Name: 5-chloro-2-hydroxy-3-{2-[(1Z)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}benzene-1-sulfonic acid
Synonyms: 5-chloro-2-hydroxy-3-{2-[(1Z)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}benzenesulfonic acid
Molecular Weight (g/mol): 344.72
Molecular Formula: C12H9ClN2O6S
InChi Key: RQIJHLQABPWAQV-ZSOIEALJSA-N
SMILES: OC1=CC(=O)C=C\C1=N\NC1=CC(Cl)=CC(=C1O)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results