missing translation for 'onlineSavingsMsg'
Learn More

Lumogallion 98.0+%, TCI America™

Catalog No. A50601G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A50601G Supplier TCI America Supplier No. A50601G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4386-25-8
Molecular Formula C12H9ClN2O6S
Molecular Weight (g/mol) 344.72
MDL Number MFCD00047770
InChI Key RQIJHLQABPWAQV-ZSOIEALJSA-N
Synonym 4-Chloro-6-(2,4-dihydroxyphenylazo)-1-phenol-2-sulfonic Acid, 2,2′C,4′C-Trihydroxy-5-chloroazobenzene-3-sulfonic Acid
PubChem CID 5772873
IUPAC Name 5-chloro-2-hydroxy-3-{2-[(1Z)-2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}benzene-1-sulfonic acid
SMILES OC1=CC(=O)C=C\C1=N\NC1=CC(Cl)=CC(=C1O)S(O)(=O)=O

Specifications

Quantity 1 g
Formula Weight 344.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lumogallion [Fluorimetric reagent for Al, Ga and other metals]
RTECSNumber DB5300000
TSCA No