accessibility menu, dialog, popup

UserName

CAS RN 4376-18-5

IUPAC Name: 2-(methoxycarbonyl)benzoic acid
Synonyms: methyl hydrogen phthalate monomethylphthalate
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: FNJSWIPFHMKRAT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)