missing translation for 'onlineSavingsMsg'
Learn More

Mono-Methyl Phthalate Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022474
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022474 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022474 Supplier Merck Emd Millipore Supplier No. 36926100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mono-Methyl phthalate belongs to the class of phthalates that is broadly employed as plasticizers in various domestic. Commonly used in industrial products, personal care products, pharmaceuticals, medical devices, and paints.

Specifications

CAS 4376-18-5
Melting Point 82°C to 84°C (literature)
Molecular Formula C9H8O4
Linear Formula 2-(HO2C)C6H4CO2CH3
MDL Number MFCD00002466
Quantity 100 mg
UN Number NONH for all modes of transport
Synonym Monomethyl phthalate
Formula Weight 180.16
Grade Analytical Standard
Physical Form Neat
Show More Show Less