accessibility menu, dialog, popup

UserName

HOOOHO

CAS RN 4355-11-7

IUPAC Name: 2-[1-(carboxymethyl)cyclohexyl]acetic acid
Synonyms: [1-(carboxymethyl)cyclohexyl]acetic acid
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: YQPCHPBGAALCRT-UHFFFAOYSA-N
SMILES: OC(=O)CC1(CC(O)=O)CCCCC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)