missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclohexanediacetic Acid 98.0+%, TCI America™

Catalog No. C144425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C144425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C144425G Supplier TCI America Supplier No. C144425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4355-11-7
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00001519
InChI Key YQPCHPBGAALCRT-UHFFFAOYSA-N
Synonym 1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
PubChem CID 78061
IUPAC Name 2-[1-(carboxymethyl)cyclohexyl]acetic acid
SMILES C1CCC(CC1)(CC(=O)O)CC(=O)O

Specifications

Melting Point 181°C
Color White
Quantity 25 g
Formula Weight 200.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Cyclohexanediacetic Acid