accessibility menu, dialog, popup

UserName

Last Chance to Save on Spotlight Special Offers

Act Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 4350-09-8

NH(S)ONH2OHHO

CAS RN 4350-09-8

IUPAC Name: (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
Synonyms: 5-hydroxy-L-tryptophan
Molecular Weight (g/mol): 220.23
Molecular Formula: C11H12N2O3
InChi Key: LDCYZAJDBXYCGN-IMWMWJONNA-N
SMILES: N[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.