accessibility menu, dialog, popup

UserName

CAS RN 4338-06-1

ONONOO

CAS RN 4338-06-1

IUPAC Name: 1-(4-nitrophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-nitrophenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 218.17
Molecular Formula: C10H6N2O4
InChi Key: CVKDEEISKBRPEQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N1C(=O)C=CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results