missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Nitrophenyl)maleimide 98.0+%, TCI America™

Catalog No. N06385G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06385G Supplier TCI America Supplier No. N06385G
Only null left

Chemical Identifiers

CAS 4338-06-1
Molecular Formula C10H6N2O4
Molecular Weight (g/mol) 218.17
MDL Number MFCD00022576
InChI Key CVKDEEISKBRPEQ-UHFFFAOYSA-N
PubChem CID 236787
IUPAC Name 1-(4-nitrophenyl)pyrrole-2,5-dione
SMILES C1=CC(=CC=C1N2C(=O)C=CC2=O)[N+](=O)[O-]

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 218.17
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Nitrophenyl)maleimide