accessibility menu, dialog, popup

UserName

CAS RN 432-60-0

H3C(S)(S)(R)(R)(S)(R)HOH2C

CAS RN 432-60-0

IUPAC Name: (1R,3aS,3bR,9aR,9bS,11aS)-11a-methyl-1-(prop-2-en-1-yl)-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
Synonyms: allylestrenol allylestrenol (substance) gestanol gestanon gestanin allilestrenolo gestanyn organon brand of allylestrenol allyloestrenol - chemical alilestrenol
Molecular Weight (g/mol): 300.49
Molecular Formula: C21H32O
InChi Key: ATXHVCQZZJYMCF-XUDSTZEESA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)CC=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results