missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allylestrenol 98.0+%, TCI America™
Supplier: TCI America A25701G
Specifications
| Allylestrenol | |
| 80°C | |
| C21H32O | |
| 1 g | |
| ATXHVCQZZJYMCF-XUDSTZEESA-N | |
| (1R,3aS,3bR,9aR,9bS,11aS)-11a-methyl-1-(prop-2-en-1-yl)-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol | |
| 235905 | |
| 300.49 | |
| Crystalline Powder |
| 432-60-0 | |
| White | |
| MFCD00198957 | |
| 17alpha-Allyl-4-estren-17beta-ol | |
| C[C@]12CC[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)CC=C | |
| 300.49 | |
| CHEBI:31189 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 432-60-0 | |
| 300.49 | |
| ATXHVCQZZJYMCF-XUDSTZEESA-N | |
| 235905 | |
| (1R,3aS,3bR,9aR,9bS,11aS)-11a-methyl-1-(prop-2-en-1-yl)-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol |
| C21H32O | |
| MFCD00198957 | |
| 17alpha-Allyl-4-estren-17beta-ol | |
| CHEBI:31189 | |
| C[C@]12CC[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)CC=C |
Safety and Handling
RTECSNumber : KG7960000
TSCA : No