CAS RN 4316-74-9
IUPAC Name:
sodium 2-(methylamino)ethane-1-sulfonate
Synonyms:
sodium methyltaurate sodium methyl cocoyl taurate
Molecular Weight (g/mol):
161.15
Molecular Formula:
C3H8NNaO3S
InChi Key:
KKDONKAYVYTWGY-UHFFFAOYSA-M
SMILES:
[Na+].CNCCS([O-])(=O)=O
1
–
1
of
1
results
1
N-Methyltaurine Sodium Salt (62-66% in Water), TCI America™
CAS: 4316-74-9 Molecular Formula: C3H8NNaO3S Molecular Weight (g/mol): 161.151 MDL Number: MFCD00066598 InChI Key: KKDONKAYVYTWGY-UHFFFAOYSA-M PubChem CID: 23684896 IUPAC Name: sodium;2-(methylamino)ethanesulfonate SMILES: CNCCS(=O)(=O)[O-].[Na+]