missing translation for 'onlineSavingsMsg'
Learn More

N-Methyltaurine Sodium Salt (62-66% in Water), TCI America™

Catalog No. M043425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M043425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M043425G Supplier TCI America Supplier No. M043425G
Only null left

Chemical Identifiers

CAS 4316-74-9
Molecular Formula C3H8NNaO3S
Molecular Weight (g/mol) 161.151
MDL Number MFCD00066598
InChI Key KKDONKAYVYTWGY-UHFFFAOYSA-M
PubChem CID 23684896
IUPAC Name sodium;2-(methylamino)ethanesulfonate
SMILES CNCCS(=O)(=O)[O-].[Na+]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 161.15
Physical Form Liquid
Chemical Name or Material N-Methyltaurine Sodium Salt (62-66% in Water)