accessibility menu, dialog, popup

UserName

H3COOCH3SH2N

CAS RN 43088-42-2

IUPAC Name: ethyl 2-amino-4-methylthiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-methylthiophene-3-carboxylate
Molecular Weight (g/mol): 185.24
Molecular Formula: C8H11NO2S
InChi Key: ILYCZKOBLRJJSW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)