missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Amino-4-methylthiophene-3-carboxylate 98.0+%, TCI America™

Catalog No. E11035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11035G 5 g
E11031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11035G Supplier TCI America Supplier No. E11035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 43088-42-2
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD00051669
InChI Key ILYCZKOBLRJJSW-UHFFFAOYSA-N
Synonym 2-amino-4-methylthiophene-3-carboxylic acid ethyl ester, 2-amino-4-methyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl2-amino-4-methylthiophene-3-carboxylate, ethyl 2-amino-4-methyl-3-thiophenecarboxylate, acmc-20a9ux, maybridge1_003755, 2-amino-4-methyl-3-ethoxycarbonyl-thiophene, ethyl 2-amino4-methylthiophene-3-carboxylate, 2-amino-3-ethoxycarbonyl-4-methylthiophene
PubChem CID 521132
IUPAC Name ethyl 2-amino-4-methylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C)N

Specifications

Melting Point 81°C
Color Yellow
Quantity 5 g
Formula Weight 185.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Amino-4-methylthiophene-3-carboxylate