CAS RN 43040-76-2
CAS RN 43040-76-2
IUPAC Name:
5-(dibutylamino)naphthalene-1-sulfonyl chloride
Synonyms:
5-(dibutylamino)naphthalene-1-sulfonyl chloride
Molecular Weight (g/mol):
353.91
Molecular Formula:
C18H24ClNO2S
InChi Key:
PIZHBGYIKYGXBV-UHFFFAOYSA-N
SMILES:
CCCCN(CCCC)C1=C2C=CC=C(C2=CC=C1)S(Cl)(=O)=O
1
–
2
of
2
results
1
Bansyl Chloride 98.0+%, TCI America™
CAS: 43040-76-2 Molecular Formula: C18H24ClNO2S Molecular Weight (g/mol): 353.91 MDL Number: MFCD00059134 InChI Key: PIZHBGYIKYGXBV-UHFFFAOYSA-N Synonym: 5-(Dibutylamino)naphthalene-1-sulfonyl Chloride PubChem CID: 191102 IUPAC Name: 5-(dibutylamino)naphthalene-1-sulfonyl chloride SMILES: CCCCN(CCCC)C1=C2C=CC=C(C2=CC=C1)S(Cl)(=O)=O
2
Bansyl Chloride (10% in Hexane), TCI America™
CAS: 43040-76-2 Molecular Formula: C18H24ClNO2S Molecular Weight (g/mol): 353.91 MDL Number: MFCD00059134 InChI Key: PIZHBGYIKYGXBV-UHFFFAOYSA-N Synonym: 5-(N,N-Dibutylamino)naphthalene-1-sulfonyl Chloride PubChem CID: 191102 IUPAC Name: 5-(dibutylamino)naphthalene-1-sulfonyl chloride SMILES: CCCCN(CCCC)C1=C2C=CC=C(C2=CC=C1)S(Cl)(=O)=O