missing translation for 'onlineSavingsMsg'
Learn More

Bansyl Chloride (10% in Hexane), TCI America™

Catalog No. B11221ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11221ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11221ML Supplier TCI America Supplier No. B11221ML
Only null left

Chemical Identifiers

CAS 43040-76-2
Molecular Formula C18H24ClNO2S
Molecular Weight (g/mol) 353.91
MDL Number MFCD00059134
InChI Key PIZHBGYIKYGXBV-UHFFFAOYSA-N
Synonym 5-(N,N-Dibutylamino)naphthalene-1-sulfonyl Chloride
PubChem CID 191102
IUPAC Name 5-(dibutylamino)naphthalene-1-sulfonyl chloride
SMILES CCCCN(CCCC)C1=C2C=CC=C(C2=CC=C1)S(Cl)(=O)=O

Specifications

Color Yellow
Quantity 1 mL
UN Number 2924
Formula Weight 353.91
Physical Form Liquid
Chemical Name or Material Bansyl Chloride (10% in Hexane)