accessibility menu, dialog, popup

UserName

NN

CAS RN 43036-06-2

IUPAC Name: 3-(1H-pyrrol-1-yl)propanenitrile
Synonyms: 3-(pyrrol-1-yl)propanenitrile
Molecular Weight (g/mol): 120.16
Molecular Formula: C7H8N2
InChi Key: IYOLJLGYJMJLSU-UHFFFAOYSA-N
SMILES: N#CCCN1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)