missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)propanenitrile, 97%, Thermo Scientific™

Catalog No. p-4812854
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD01949DA 1g
CD01949EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CD01949DA Supplier Thermo Scientific Chemicals Supplier No. CD01949DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 43036-06-2
Molecular Formula C7H8N2
Molecular Weight (g/mol) 120.16
MDL Number MFCD00003092
InChI Key IYOLJLGYJMJLSU-UHFFFAOYSA-N
Synonym 3-1h-pyrrol-1-yl propanenitrile, n-2-cyanoethyl pyrrole, 1-2-cyanoethyl pyrrole, pyrrole-1-propionitrile, 1h-pyrrole-1-propanenitrile, 3-pyrrol-1-yl propiononitrile, n-2-cyanoethyl-pyrrole, 3-pyrrol-1-yl propanenitrile, n-cyanoethylpyrrole, n-2-cyanoethyl pyrole
PubChem CID 96527
IUPAC Name 3-pyrrol-1-ylpropanenitrile
SMILES N#CCCN1C=CC=C1

Specifications

Assay Percent Range 97%
Quantity 1g
Formula Weight 120.15
Chemical Name or Material 3-(1H-Pyrrol-1-yl)propanenitrile