accessibility menu, dialog, popup

UserName

CAS RN 42926-52-3

CH3OClO

CAS RN 42926-52-3

IUPAC Name: 2-ethoxybenzoyl chloride
Synonyms: 2-ethoxybenzoyl chloride
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: MDKAAWDKKBFSTK-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results