missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzoyl Chloride 98.0+%, TCI America™

Catalog No. E067625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E067625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E067625G Supplier TCI America Supplier No. E067625G
Only null left

Chemical Identifiers

CAS 42926-52-3
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00016348
InChI Key MDKAAWDKKBFSTK-UHFFFAOYSA-N
Synonym o-ethoxybenzoyl chloride, benzoyl chloride, 2-ethoxy, 2-ethoxybenzoylchloride, acmc-209jrt, 2-ethoxy-benzoyl chloride, benzoyl chloride,2-ethoxy, 2-ethoxybenzoic acid chloride, ksc491c5l, 2-ethoxybenzoyl chloride
PubChem CID 3016397
IUPAC Name 2-ethoxybenzoyl chloride
SMILES CCOC1=CC=CC=C1C(=O)Cl

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 g
UN Number 3265
Formula Weight 184.62
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzoyl Chloride