accessibility menu, dialog, popup

UserName

CH3NH3CH3CH3CFPFFFFF

CAS RN 429-07-2

IUPAC Name: hexafluoro-λ⁵-phosphanuide; tetraethylazanium
Synonyms: tetraethylammonium hexafluorophosphate
Molecular Weight (g/mol): 275.22
Molecular Formula: C8H20F6NP
InChi Key: KLKUOIXSIDDDCN-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CC[N+](CC)(CC)CC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results