missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hexafluorophosphate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014185
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014185 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014185 Supplier Merck Emd Millipore Supplier No. 8662525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

CAS 429-07-2
Molecular Weight (g/mol) 275.22
MDL Number MFCD00043179

Specifications

Melting Point ≥300°C
Linear Formula (C2H5)4 N(PF6)
Quantity 25 g
Solubility Information Acetonitrile: 0.1 g/mL, clear, colorless
Formula Weight 275.22
Percent Purity ≥99.0% (T); ≥99.0%
Grade For electrochemical analysis
Physical Form Crystals
RTECSNumber BS6500000