accessibility menu, dialog, popup

UserName

CAS RN 42564-51-2

IUPAC Name: 4-fluoro-3-nitrobenzaldehyde
Synonyms: 4-fluoro-3-nitrobenzaldehyde
Molecular Weight (g/mol): 169.11
Molecular Formula: C7H4FNO3
InChi Key: ILKWFRCNNILIJW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(C=O)=CC=C1F
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)