missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. AAH2618106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2618106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2618106 Supplier Thermo Scientific Chemicals Supplier No. H2618106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42564-51-2 | Molecular Formula: C7H4FNO3 | Molecular Weight: 169.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42564-51-2
Molecular Formula C7H4FNO3
Molecular Weight (g/mol) 169.111
MDL Number MFCD01861388
InChI Key ILKWFRCNNILIJW-UHFFFAOYSA-N
Synonym 3-nitro-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzaldehyde, pubchem22572, acmc-1ao2q, 3-nitro-4 fluorobenzaldehyde, ksc497m9r, 2-fluoro-5-formylnitrobenzene, #, 4-fluoranyl-3-nitro-benzaldehyde
PubChem CID 598129
IUPAC Name 4-fluoro-3-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])F

Specifications

Melting Point 44°C to 47°C
Flash Point >110°C (230°F)
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 169.11
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-nitrobenzaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only