accessibility menu, dialog, popup

UserName

CAS RN 4238-71-5

IUPAC Name: 1-benzyl-1H-imidazole
Synonyms: 1-benzylimidazole|BIM
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: KKKDZZRICRFGSD-UHFFFAOYSA-N
SMILES: C(N1C=CN=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)