CAS RN 42270-37-1
CAS RN 42270-37-1
IUPAC Name:
1-(1,3-thiazol-2-yl)piperazine
Synonyms:
piperazine, 1-(2-thiazolyl)-
Molecular Weight (g/mol):
169.25
Molecular Formula:
C7H11N3S
InChi Key:
WQFWIVTXNKRNJZ-UHFFFAOYSA-N
SMILES:
C1CN(CCN1)C1=NC=CS1
1
–
1
of
1
results
1
1-(2-Thiazolyl)piperazine 98.0+%, TCI America™
CAS: 42270-37-1 Molecular Formula: C7H11N3S Molecular Weight (g/mol): 169.25 MDL Number: MFCD00778419 InChI Key: WQFWIVTXNKRNJZ-UHFFFAOYSA-N Synonym: 2-(1-Piperazinyl)thiazole PubChem CID: 911806 IUPAC Name: 1-(1,3-thiazol-2-yl)piperazine SMILES: C1CN(CCN1)C1=NC=CS1