accessibility menu, dialog, popup

UserName

CH3OOOH(E/Z)

CAS RN 42220-77-9

IUPAC Name: 1-(2-hydroxyphenyl)-3-(2-methoxyphenyl)prop-2-en-1-one
Synonyms: 1-(2-hydroxyphenyl)-3-(2-methoxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol): 254.29
Molecular Formula: C16H14O3
InChi Key: SNTIPKTZVAKPOX-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C=CC(=O)C1=CC=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results