missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-2-methoxychalcone 98.0+%, TCI America™

Catalog No. H14931G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H14931G Supplier TCI America Supplier No. H14931G
Only null left

Chemical Identifiers

CAS 42220-77-9
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00016443
InChI Key SNTIPKTZVAKPOX-ZHACJKMWSA-N
Synonym 2-(2-Methoxybenzylidene)-2′C-hydroxyacetophenone
PubChem CID 5709142
IUPAC Name (E)-1-(2-hydroxyphenyl)-3-(2-methoxyphenyl)prop-2-en-1-one
SMILES COC1=CC=CC=C1C=CC(=O)C2=CC=CC=C2O

Specifications

Melting Point 111°C
Quantity 1 g
Formula Weight 254.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-2-methoxychalcone