accessibility menu, dialog, popup

UserName

CAS RN 422-85-5

FFFFFFFBr

CAS RN 422-85-5

IUPAC Name: 1-bromo-1,1,2,2,3,3,3-heptafluoropropane
Synonyms: 1-bromo-1,1,2,2,3,3,3-heptafluoropropane
Molecular Weight (g/mol): 248.93
Molecular Formula: C3BrF7
InChi Key: LANNRYWUUQMNPF-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results