accessibility menu, dialog, popup

UserName

NH2NNOO

CAS RN 4214-76-0

IUPAC Name: 5-nitropyridin-2-amine
Synonyms: 2-pyridinamine, 5-nitro-
Molecular Weight (g/mol): 139.11
Molecular Formula: C5H5N3O2
InChi Key: UGSBCCAHDVCHGI-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=N1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (6)