missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitropyridine, 99%

Catalog No. AC104232500
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104232500 250 g Glass Bottle
AC104230050 5 g Glass Bottle
AC104230250 25 g Glass Bottle
AC104231000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC104232500 Supplier Thermo Scientific Chemicals Supplier No. 104232500

CAS: 4214-76-0 | C5H5N3O2 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4214-76-0
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006325
InChI Key UGSBCCAHDVCHGI-UHFFFAOYSA-N
Synonym 2-amino-5-nitropyridine, 2-pyridinamine, 5-nitro, 5-nitro-2-aminopyridine, 5-nitro-2-pyridinamine, 2-amino-5-nitro pyridine, pyridine, 2-amino-5-nitro, 5-nitro-pyridin-2-ylamine, 5-nitro-pyridin-2-yl amine, 5-nitro-2-pyridylamine, 5-nitropyridin-2-amin
PubChem CID 77888
IUPAC Name 5-nitropyridin-2-amine
SMILES NC1=CC=C(C=N1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 186.0°C to 190.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula NH2(C5H3N)NO2
Quantity 250 g
Beilstein 22,I,631
Solubility Information Solubility in water: 1.6g/L water (13°C).
Formula Weight 139.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitropyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 224-145-6

RUO – Research Use Only