accessibility menu, dialog, popup

UserName

CAS RN 42137-88-2

IUPAC Name: N,N-bis(2-chloroethyl)-4-methylbenzene-1-sulfonamide
Synonyms: N,N-bis(2-chloroethyl)-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 296.21
Molecular Formula: C11H15Cl2NO2S
InChi Key: PTVBBIMKLOMGSY-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N(CCCl)CCCl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)