missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-chloroethyl)-p-toluenesulfonamide, tech. 90%

Catalog No. AAL0529614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0529614 25 g
AAL0529606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0529614 Supplier Thermo Scientific Chemicals Supplier No. L0529614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42137-88-2 | C11H15Cl2NO2S | 296.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42137-88-2
Molecular Formula C11H15Cl2NO2S
Molecular Weight (g/mol) 296.206
MDL Number MFCD00018944
InChI Key PTVBBIMKLOMGSY-UHFFFAOYSA-N
Synonym n,n-bis 2-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-p-toluenesulphonamide, n,n-bis .beta.-chloroethyl-p-toluenesulfonamide, n,n-bis 2-chloroethyl-4-toluenesulfonamide, n,n-bis 2'-chloroethyl-4-methylbenzenesulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulfonamide, n,n-bis 2-chloroethyl-4-methylbenzene sulphonamide, benzenesulfonamide, n,n-bis 2-chloroethyl-4-methyl, n,n-bis-2-chloro-ethyl-4-methyl-benzenesulfonamide
PubChem CID 96039
IUPAC Name N,N-bis(2-chloroethyl)-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(CCCl)CCCl

Specifications

Melting Point 45°C to 47°C
Quantity 25 g
Beilstein 2134258
Formula Weight 296.22
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N-Bis(2-chloroethyl)-p-toluenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-677-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only