accessibility menu, dialog, popup

UserName

CAS RN 4209-90-9

IUPAC Name: 2,2-dimethylhexan-3-ol
Synonyms: 3-hexanol, 2,2-dimethyl-
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: PFHLGQKVKALLMD-UHFFFAOYNA-N
SMILES: CCCC(O)C(C)(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)