missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-3-hexanol 98.0+%, TCI America™

Catalog No. D153210ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D153210ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D153210ML Supplier TCI America Supplier No. D153210ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4209-90-9
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00021929
InChI Key PFHLGQKVKALLMD-UHFFFAOYSA-N
Synonym 2,2-dimethyl-3-hexanol, 3-hexanol, 2,2-dimethyl, acmc-209jni, 3-hexanol,2-dimethyl, n-propyl-tert-butylcarbinoll, pfhlgqkvkallmd-uhfffaoysa
PubChem CID 98265
IUPAC Name 2,2-dimethylhexan-3-ol
SMILES CCCC(C(C)(C)C)O

Specifications

Color Colorless
Boiling Point 156°C
Quantity 10 mL
UN Number 1987
Formula Weight 130.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethyl-3-hexanol
EINECSNumber (2)-0217
TSCA No