CAS RN 42017-89-0
IUPAC Name:
2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
Synonyms:
fenofibric acid
Molecular Weight (g/mol):
318.75
Molecular Formula:
C17H15ClO4
InChi Key:
MQOBSOSZFYZQOK-UHFFFAOYSA-N
SMILES:
CC(C)(OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1)C(O)=O
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific™ Fenofibric acid
Thermo Scientific™ Fenofibric acid
CAS: 42017-89-0 Chemical Name or Material: Fenofibric acid
2
Fenofibric Acid 98.0+%, TCI America™
CAS: 42017-89-0 Molecular Formula: C17H15ClO4 Molecular Weight (g/mol): 318.75 MDL Number: MFCD00792461 InChI Key: MQOBSOSZFYZQOK-UHFFFAOYSA-N Synonym: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid PubChem CID: 64929 ChEBI: CHEBI:83469 IUPAC Name: 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid SMILES: CC(C)(OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1)C(O)=O