missing translation for 'onlineSavingsMsg'
Learn More

Fenofibric Acid 98.0+%, TCI America™

Catalog No. F10115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10115G 5 g
F10111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10115G Supplier TCI America Supplier No. F10115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42017-89-0
Molecular Formula C17H15ClO4
Molecular Weight (g/mol) 318.75
MDL Number MFCD00792461
InChI Key MQOBSOSZFYZQOK-UHFFFAOYSA-N
Synonym 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid
PubChem CID 64929
ChEBI CHEBI:83469
IUPAC Name 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
SMILES CC(C)(OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point 181°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 318.75
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fenofibric Acid
RTECSNumber UA2453000
TSCA No