accessibility menu, dialog, popup

UserName

H2CFFBr

CAS RN 420-90-6

IUPAC Name: 3-bromo-3,3-difluoroprop-1-ene
Synonyms: 3-bromo-3,3-difluoroprop-1-ene
Molecular Weight (g/mol): 156.96
Molecular Formula: C3H3BrF2
InChi Key: GDDNTTHUKVNJRA-UHFFFAOYSA-N
SMILES: FC(F)(Br)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results