CAS RN 41830-81-3
IUPAC Name:
N-ethyl-3-(ethylimino)-2,8-dimethyl-3H-phenoxazin-7-amine
Synonyms:
N-ethyl-7-(ethylimino)-2,8-dimethylphenoxazin-3-amine
Molecular Weight (g/mol):
295.39
Molecular Formula:
C18H21N3O
InChi Key:
PWLZRLVLUJPWOB-UHFFFAOYSA-N
SMILES:
CCNC1=C(C)C=C2N=C3C=C(C)C(C=C3OC2=C1)=NCC
1
–
1
of
1
results
1
Oxazine 4 Perchlorate 98.0+%, TCI America™
CAS: 41830-81-3 Molecular Formula: C18H21N3O Molecular Weight (g/mol): 295.39 MDL Number: MFCD00042008 InChI Key: PWLZRLVLUJPWOB-UHFFFAOYSA-N Synonym: oxazine 4 perchlorate,7e-n-ethyl-7-ethylimino-2,8-dimethylphenoxazin-3-amine; perchloric acid PubChem CID: 90472504 IUPAC Name: N-ethyl-3-(ethylimino)-2,8-dimethyl-3H-phenoxazin-7-amine SMILES: CCNC1=C(C)C=C2N=C3C=C(C)C(C=C3OC2=C1)=NCC