missing translation for 'onlineSavingsMsg'
Learn More

Oxazine 4 Perchlorate 98.0+%, TCI America™

Catalog No. O0479200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0479200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0479200MG Supplier TCI America Supplier No. O0479200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41830-81-3
Molecular Formula C18H21N3O
Molecular Weight (g/mol) 295.39
MDL Number MFCD00042008
InChI Key PWLZRLVLUJPWOB-UHFFFAOYSA-N
Synonym oxazine 4 perchlorate, 7e-n-ethyl-7-ethylimino-2,8-dimethylphenoxazin-3-amine; perchloric acid
PubChem CID 90472504
IUPAC Name N-ethyl-3-(ethylimino)-2,8-dimethyl-3H-phenoxazin-7-amine
SMILES CCNC1=C(C)C=C2N=C3C=C(C)C(C=C3OC2=C1)=NCC

Specifications

Quantity 200 mg
Formula Weight 395.84
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Oxazine 4 Perchlorate