accessibility menu, dialog, popup

UserName

H3CHOO

CAS RN 4169-04-4

IUPAC Name: 2-phenoxypropan-1-ol
Synonyms: propylene glycol monophenyl ether
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: LOJHHQNEBFCTQK-UHFFFAOYNA-N
SMILES: CC(CO)OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results