missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxypropanol 96.0+%, TCI America™

Catalog No. P11115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11115G 5 g
P111125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11115G Supplier TCI America Supplier No. P11115G
Only null left

Chemical Identifiers

CAS 4169-04-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00142958
InChI Key LOJHHQNEBFCTQK-UHFFFAOYNA-N
Synonym Propylene Glycol 2-Monophenyl Ether
PubChem CID 20129
IUPAC Name 2-phenoxypropan-1-ol
SMILES CC(CO)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 244°C
Quantity 5 g
Formula Weight 152.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxypropanol
EINECSNumber (3)-0560
RTECSNumber UB8886000
TSCA Yes